Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVPL Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP259099 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259099 Supplier Novus Biologicals Supplier No. NBP259099

Rabbit Polyclonal Antibody

EVPL Polyclonal specifically detects EVPL in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen EVPL
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunoprecipitation
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias 210 KDa Cornified Envelope Precursor Protein, 210 KDa Paraneoplastic Pemphigus Antigen, envoplakin, EVPK, p210
Gene Symbols EVPL
Host Species Rabbit
Immunogen Maps to a region between 1983 and 2033
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2125
Target Species Human
Content And Storage Store at 4C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.