Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EWSR1 Antibody (5H7), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB361923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361923 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361923 Supplier Novus Biologicals Supplier No. NBP192686MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EWSR1 Monoclonal antibody specifically detects EWSR1 in Human, Mouse, Rat, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen EWSR1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 5H7
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS
Host Species Mouse
Immunogen Full length recombinant human EWSR1 expressed in and purified from E. coli. [UniProt# Q01844]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2130
Target Species Human, Mouse, Rat, Canine, Equine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.