Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EWSR1 Antibody (5H7), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$207.00 - $499.50

Specifications

Antigen EWSR1
Clone 5H7
Concentration 1 mg/mL
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP192686SS
SDP
View Documents
Novus Biologicals
NBP192686SS
0.025 mL
Each for $207.00
Only null left
Add to Cart
 
NBP192686
SDP
View Documents
Novus Biologicals
NBP192686
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

EWSR1 Monoclonal specifically detects EWSR1 in Human, Mouse, Rat, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

EWSR1
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Cancer, Cellular Markers, Neuroscience, Tumor Suppressors
50% PBS, 50% glycerol with 5mM Sodium Azide
bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS
EWSR1
IgG2b
Protein G purified
80 kDa
5H7
Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
Monoclonal
Purified
RUO
Human, Mouse, Rat, Canine, Equine
Q01844
2130
Full length recombinant human EWSR1 expressed in and purified from E. coli. [UniProt# Q01844]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.