Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EWSR1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10075256 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10075256 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10075256 Supplier Novus Biologicals Supplier No. NB10075256
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

EWSR1 Polyclonal specifically detects EWSR1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen EWSR1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q01844
Gene Alias bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS
Gene Symbols EWSR1
Host Species Chicken
Immunogen The immunogen for this antibody was a sequence from 380a.a. to 448a.a. from RNA-binding protein EWSR1(a 656a.a. long protein)
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2130
Test Specificity RNA-binding protein EWSR1
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.