Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Stainless Steel Adson Dissecting Forcep

Catalog No. 18100912 Shop All Excelta Corporation Products
Click to view available options
Length (Metric):
120 mm

Stainless Steel Adson Dissecting Forcep ideal for dissecting, with long very slender nose.

  • Ideal for dissection

Specifications

Product Type Adson Dissecting Forcep
Autoclavable Autoclavable
Color Silver
Material Stainless Steel (AISI 440)
For Use With (Application) Dissecting
Length (English) 4.75 in.
Length (Metric) 120 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.