Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YSI™ Exchangeable Unit with Calibration Certificate
Shop All Yellow Springs Instrument (YSI) Products
Click to view available options
Volume (Metric) Sample:
10 mL
20 mL
50 mL
Description
Intelligent exchangeable heads for the piston burette TITRONIC™ 500 and the titrators TitroLine™ 7000, 7500, 7750 und 7800.
- Size options of 5, 10, 20 und 50 ml
- Compact, space saving footprint
- Easy to exchange
- All relevant reagent and unit data are stored in the integrated RFID-chip including:
- Burette size (ml)
- Reagent/titrant name
- Titrant concentration or titer value of solution
- Date of manufacture or expiration date of the reagent.
ISO 8655 Certified (“C” certificate included)
Scope of delivery:
5, 10 , 20 or 50 ml burette, with integrated chip for reagent data, with 1 L brown glass bottle for titrant, GL 45 and S 40-bottle adapter, tubes, drip tube and titration tip

Specifications
Specifications
Type | Intelligent Exchangeable Unit for TitroLine 7000+ and TITRONIC 500 |
Volume (Metric) Sample | 50 mL |
For Use With (Equipment) | TitroLine 7000+ and TITRONIC 500 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction