Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXD1 Antibody (OTI5B5), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP272443V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272443V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272443V Supplier Novus Biologicals Supplier No. NBP272443V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EXD1 Monoclonal specifically detects EXD1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen EXD1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5B5
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EXDL1, exonuclease 3′-5′ domain containing 1, exonuclease 3′-5′ domain-containing protein 1, exonuclease 3′-5′ domain-like 1, Exonuclease 3′-5′ domain-like-containing protein 1, MGC33637
Gene Symbols EXD1
Host Species Mouse
Immunogen Full length recombinant protein of human EXD1 (NP_689809) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 161829.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.