Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exendin-4 Antibody (20), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP22377A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22377A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22377A30 Supplier Novus Biologicals Supplier No. NBP223577AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Exendin-4 Monoclonal specifically detects Exendin-4 in Reptile samples. It is validated for ELISA.

Specifications

Antigen Exendin-4
Applications ELISA
Classification Monoclonal
Clone 20
Conjugate Alexa Fluor 350
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Exendin4
Host Species Mouse
Immunogen Synthetic exendin-4 adsorbed onto aluminum hydroxide gel
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Specific for exendin-4. No cross-reactivity with GLP-1, GLP-2 (human) or glucagon coated on ELISA wells
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.