Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

exo-Tetrahydrodicyclopentadiene 94.0+%, TCI America™
SDP

Catalog No. T199425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T199425G 25 g
T19945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T199425G Supplier TCI America Supplier No. T199425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2825-82-3
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD01714931
InChI Key LPSXSORODABQKT-UHFFFAOYNA-N
Synonym exo-Tricyclo[5.2.1.0(2,6)]decane
PubChem CID 11159354
IUPAC Name tricyclo[5.2.1.0²,⁶]decane
SMILES C1CC2C3CCC(C3)C2C1

Specifications

Color Yellow
Quantity 25 g
UN Number 1993
Formula Weight 136.24
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material exo-Tetrahydrodicyclopentadiene
EINECSNumber (4)-0655
RTECSNumber PB9600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.