Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOC5 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB325241 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325241 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325241 Supplier Novus Biologicals Supplier No. NBP299472PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EXOC5 Polyclonal antibody specifically detects EXOC5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen EXOC5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias exocyst complex component 5, Exocyst complex component Sec10, HSEC10, PRO1912, SEC10 (S. cerevisiae)-like 1, SEC10DKFZp666H126, SEC10L1, SEC10-like 1, SEC10-like 1 (S. cerevisiae), SEC10P
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human EXOC5.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10640
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.