Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EXOC6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | EXOC6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15543020
![]() |
Novus Biologicals
NBP15543020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155430
![]() |
Novus Biologicals
NBP155430 |
100 μL |
Each for $487.50
|
|
|||||
Description
EXOC6 Polyclonal specifically detects EXOC6 in Human, Mouse samples. It is validated for Western Blot.Specifications
EXOC6 | |
Polyclonal | |
Rabbit | |
B3KXY5 | |
54536 | |
Synthetic peptides corresponding to EXOC6(exocyst complex component 6) Antibody(against the N terminal of EXOC6. Peptide sequence MLEEETDQTYENVLAEIQSFELPVEATLRSVYDDQPNAHKKFMEKLDACI. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DKFZp761I2124, EXOC6A, exocyst complex component 6, Exocyst complex component Sec15A, FLJ1125, FLJ11251, MGC33397, SEC15A, SEC15L1SEC15-like 1 (S. cerevisiae), SEC15L3, SEC15-like 1, SEC15-like protein 1, SEC15-like protein 3, SEC15LSEC15, Sec15p | |
EXOC6 | |
IgG | |
88 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title