Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exonuclease 1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093394 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093394 Supplier Novus Biologicals Supplier No. NB10093394
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Exonuclease 1 Polyclonal specifically detects Exonuclease 1 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen Exonuclease 1
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.2 - 0.5 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q9UQ84
Gene Alias EXOI, exonuclease 1, Exonuclease I, HEX1rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1, hExo1, hExoIEC 3.1
Gene Symbols EXO1
Host Species Goat
Immunogen Peptide with sequence C-HRNYSPRPESGT corresponding to internal region according to NP_003677.3, NP_006018.3, NP_569082.1.
Molecular Weight of Antigen 94 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Core ESC Like Genes, DNA Repair, Editing and Processing Endonucleases, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9156
Test Specificity This antibody is expected to recognize all three reported isoforms (NP_003677.3; NP_006018.3 and NP_569082.1)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.