Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human EXOSC3 His Protein
SDP

Catalog No. p-7104614 Shop All Novus Biologicals Products
Click to view available options
Quantity:
0.01 mg
0.05 mg

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-275 of Human EXOSC3 The Recombinant Human EXOSC3 Protein is derived from E. coli. The Recombinant Human EXOSC3 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5 containing 0.15M NaCl, 30% glycerol, 1mM DTT
Gene ID (Entrez) 51010
Molecular Weight (g/mol) TMW: 32kDa
Quantity 0.01 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias bA3J10.7, CGI-102, exosome complex exonuclease RRP40, exosome component 3MGC15120, exosome component Rrp40, hRrp-40, hRrp40p, p10Rrp40p, Ribosomal RNA-processing protein 40, RP11-3J10.8, RRP40MGC723
Common Name EXOSC3
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.