Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosome component 4 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB047902 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047902 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047902 Supplier Novus Biologicals Supplier No. NBP297799R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Exosome component 4 Polyclonal specifically detects Exosome component 4 in Human samples. It is validated for Western Blot.

Specifications

Antigen Exosome component 4
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias exosome complex exonuclease RRP41, exosome component 4Rrp41p, exosome component Rrp41, FLJ20591, p12ARibosomal RNA-processing protein 41, RRP41A, RRP41Ski6p, SKI6hRrp41p
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Exosome component 4 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 54512
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.