Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosome component 5 Antibody (V80P1C10*C), Biotin, Novus Biologicals™
SDP

Catalog No. NBP250173B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250173B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250173B Supplier Novus Biologicals Supplier No. NBP250173B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Exosome component 5 Monoclonal specifically detects Exosome component 5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.

Specifications

Antigen Exosome component 5
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V80P1C10*C
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CML28, exosome complex exonuclease RRP46, exosome component 5Chronic myelogenous leukemia tumor antigen 28, exosome component Rrp46, hRrp46p, MGC12901, p12BMGC111224, RRP41B, Rrp46p, RRP46Ribosomal RNA-processing protein 46
Gene Symbols EXOSC5
Host Species Mouse
Immunogen Peptide Sequence - KLLMSSTKGLY
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 56915
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.