Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosome component 7 Antibody (OTI1G8), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N271722PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271722PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271722PEC7 Supplier Novus Biologicals Supplier No. NBP271722PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Exosome component 7 Monoclonal specifically detects Exosome component 7 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen Exosome component 7
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1G8
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EAP1, exosome complex exonuclease RRP42, exosome component 7Rrp42p, KIAA0116hRrp42p, p8FLJ26543, RRP42Ribosomal RNA-processing protein 42
Gene Symbols EXOSC7
Host Species Mouse
Immunogen Full length recombinant protein of human Exosome component 7 (NP_055819) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23016.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.