Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosome component 8 Antibody, Novus Biologicals™
SDP

Catalog No. NB435815 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435815 25 μL
NBP247465 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435815 Supplier Novus Biologicals Supplier No. NBP24746525UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Exosome component 8 Polyclonal specifically detects Exosome component 8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Exosome component 8
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias bA421P11.3, CBP-interacting protein 3, CIP3, EAP2, exosome complex exonuclease RRP43, exosome component 8Opa-interacting protein 2, OIP2Ribosomal RNA-processing protein 43, Opa interacting protein 2, p9RP11-421P11.3, RRP43OIP-2, Rrp43p
Gene Symbols EXOSC8
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: ALAEVNLKKKSYLNIRTHPVATSFAVFDDTLLIVDPTGEEEHLATGTLTIVMDEEGKLCCLHKPGGSGLTGAKLQDCMSRAVTRHKEVKKLMDEVIKSMK
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 11340
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.