Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosome Component 9 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP171702A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP171702A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP171702A30 Supplier Novus Biologicals Supplier No. NBP171702AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Exosome Component 9 Polyclonal specifically detects Exosome Component 9 in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Exosome Component 9
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Autoantigen PM/Scl 1, EC 3.1.13, exosome component 9Polymyositis/scleroderma autoantigen 75 kDa, p6, PM/Scl-75P75 polymyositis-scleroderma overlap syndrome-associated autoantigen, PMSCL175kD, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa
Gene Symbols EXOSC9
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of human Exosome Component 9 (within residues 250-439). [Swiss-Prot Q06265]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5393
Target Species Human, Mouse, Chicken
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.