Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosome Component 9 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP171702SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171702SS 0.025 mL
NBP171702 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP171702SS Supplier Novus Biologicals Supplier No. NBP171702SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Exosome Component 9 Polyclonal specifically detects Exosome Component 9 in Human, Mouse, Chicken samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Exosome Component 9
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000, Simple Western 1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 27543448), Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 1:50-1:100, Chromatin Immunoprecipitation (ChIP)
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. Q06265, Q9JHI7
Gene Alias Autoantigen PM/Scl 1, EC 3.1.13, exosome component 9Polymyositis/scleroderma autoantigen 75 kDa, p6, PM/Scl-75P75 polymyositis-scleroderma overlap syndrome-associated autoantigen, PMSCL175kD, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa
Gene Symbols EXOSC9
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of human Exosome Component 9 (within residues 250-439). [Swiss-Prot Q06265]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5393
Test Specificity Human, Mouse and Chicken. Immunogen has 83% identity to rat and 90% identity to bovine.
Target Species Human, Mouse, Chicken
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.