Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosome Component 9 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB338160 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338160 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338160 Supplier Novus Biologicals Supplier No. NBP171702MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Exosome Component 9 Polyclonal antibody specifically detects Exosome Component 9 in Human, Mouse, Chicken samples. It is validated for Western Blot, Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)

Specifications

Antigen Exosome Component 9
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP)
Formulation 50mM Sodium Borate
Gene Alias Autoantigen PM/Scl 1, EC 3.1.13, exosome component 9Polymyositis/scleroderma autoantigen 75 kDa, p6, PM/Scl-75P75 polymyositis-scleroderma overlap syndrome-associated autoantigen, PMSCL175kD, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of human Exosome Component 9 (within residues 250-439). [Swiss-Prot Q06265]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5393
Target Species Human, Mouse, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.