Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exostosin-like 2/EXTL2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216394 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216394 Supplier Novus Biologicals Supplier No. NBP216394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Exostosin-like 2/EXTL2 Polyclonal specifically detects Exostosin-like 2/EXTL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Exostosin-like 2/EXTL2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UBQ6
Gene Alias Alpha-1,4-N-acetylhexosaminyltransferase EXTL2, alpha-1,4-N-acteylhexosaminyltransferase, Alpha-GalNAcT EXTL2, EC 2.4.1.223, exostoses (multiple)-like 2, exostosin-like 2, EXTR2, EXT-related protein 2, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, processed exostosin-like 2
Gene Symbols EXTL2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EXTL2. The exact sequence is proprietary.
Molecular Weight of Antigen 37 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2135
Test Specificity Human. Expected to cross react based on sequence identity: mouse (87%), Rat (85%), cow (87%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.