Learn More
Description
Specifications
Specifications
| Antigen | Ext2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.57mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000 |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal |
| Gene Alias | EC 2.4.1.224, EC 2.4.1.225, exostoses (multiple) 2, exostosin 2, exostosin-2, Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan4-alpha-N-acetylglucosaminyltransferase, Multiple exostoses protein 2, N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase, Putative tumor suppressor protein EXT2, SOTV |
| Gene Symbols | EXT2 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
