Learn More
Description
- Finger-tight design for wrench-free use
- Available for Standard (1/16 in.) and Compact (Agilent-style) ferrules
- Made from high-quality stainless steel
- Meets or exceeds original manufacturer's performance
Specifications
Specifications
| Type | Extended Capillary Column Nut Kit |
| For Use With (Equipment) | Standard Ferrules |
| Material | Stainless Steel |
| Color | Silver |
| Description | Finger-Tight Design |
| For Use With (Application) | Agilent GCs |
| Quantity | 1 Kit |
| Includes | Extended Reducing Nut and Extended Capillary Column Nut for Standard Ferrules |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
