Learn More
Description
- Finger-tight design for wrench-free use
- Available for Standard (1/16 in.) and Compact (Agilent-style) ferrules
- Made from high-quality stainless steel
- Meets or exceeds original manufacturer's performance
Specifications
Specifications
| For Use With (Equipment) | Compact Ferrules |
| Color | Silver |
| For Use With (Application) | Agilent GCs |
| Description | Finger-Tight Design |
| Material | Stainless Steel |
| Type | Extended Capillary Column Nut Kit |
| Quantity | 1 Kit |
| Includes | Extended Reducing Nut and Extended Capillary Column Nut for Compact Ferrules |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
