Learn More
Air Impurities Removal Systems Extract-ALL™ Filters for Source Capture Air Cleaning Systems
SDP
$121.67 - $665.28
Specifications
Product Type Filter
View More Specs
Products 2
Catalog Number Mfr. No. Efficiency Filter Type For Use With (Application) For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Efficiency Filter Type For Use With (Application) For Use With (Equipment) Price Quantity & Availability  
19-097-430
SDP
Encompass
View Documents
Air Impurities Removal Systems
F9814A
0.9997 HEPA Capture soldering fumes and vapors containing lead, (final adsorption carbon module) captures volatile solvents, paint fumes and other noxious odors Air cleaning systems
Each for $665.28
Only null left
Add to Cart
19-097-433
SDP
Encompass
View Documents
Air Impurities Removal Systems
RF9812A
0.6 Carbon Captures solvent, touch-up paint, spray glue fumes and other gaseous odors F-981-4A source capture air cleaning system HEPA filter
Each for $121.67
Only null left
Add to Cart
Description
Description
Specifications
Specifications
Filter
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.