Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EYA1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146136 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146136 Supplier Novus Biologicals Supplier No. NBP146136
Add to Cart
Add to Cart

Goat Polyclonal Antibody

EYA1 Polyclonal specifically detects EYA1 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen EYA1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q99502
Gene Alias BOP, BOR, EC 3.1.3.48, eyes absent (Drosophila) homolog 1, eyes absent homolog 1, eyes absent homolog 1 (Drosophila), MGC141875
Gene Symbols EYA1
Host Species Goat
Immunogen Peptide with sequence C-TDPTAEYSTIHSP corresponding to internal region according to NP_000494.2, NP_742056.1.
Molecular Weight of Antigen 64.593 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 2138
Test Specificity This antibody is expected to recognize all isoforms (NP_742057.1; NP_000494.2; NP_742056.1). Reported variants represent identical protein: NP_000494.2, NP_742055.1
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.