Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EYA4 Antibody, Novus Biologicals™
SDP

Catalog No. NB094207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB094207 Supplier Novus Biologicals Supplier No. NBP304071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EYA4 Polyclonal specifically detects EYA4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EYA4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:20-1:50, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias CMD1J, DFNA10, dJ78N10.1 (eyes absent), EC 3.1.3.48, eyes absent (Drosophila) homolog 4, eyes absent homolog 4, eyes absent homolog 4 (Drosophila)
Host Species Rabbit
Immunogen A synthetic Peptide of human EYA4
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 2070
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.