Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ EZ-Accu Shot™ Starved Cells

Microbiologics™ EZ-Accu Shot™ Starved Cells simplify and accelerate test procedures for growth promotion of R2A agar
$543.00
Specifications
Biosafety Level | 1 |
---|---|
Content And Storage | 2°C to 8°C |
Format | EZ-Accu Shot Starved Cells |
Manufacturing Origin | United States |
Product Type | Quality Control |
Catalog Number | Mfr. No. | Species | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Species | Price | Quantity & Availability | |||||
23-004-002
![]() |
Microbiologics™
01110SC |
Methylobacterium extorquens |
Each for $543.00
|
|
|||||
23-004-003
![]() |
Microbiologics™
0524SC |
Pseudomonas protegens |
Each for $543.00
|
|
|||||
Description
- Laboratories following Japanese Pharmacopeia (JP) guidelines know the challenges of performing Media Growth Promotion Testing on R2A agar
- The JP states that R2A media should be challenged with strains that have been starved for 3 days prior to conducting the test
- To simplify and accelerate test procedures for growth promotion of R2A agar, we have developed EZ-Accu Shot™ Starved Cells
- These test-ready control strains have been starved prior to lyophilization and deliver the required concentration of 50-200 CFU per 1 mL inoculum on R2A media
- Designed for laboratories performing the Growth Promotion Test of R2A media according to the Japanese Pharmacopeia (JP)
- No dilutions required
- Kit includes: 5 vials of a single, quantitative microorganism (1 lyophilized pellet per vial)
Specifications
1 | |
EZ-Accu Shot Starved Cells | |
Quality Control | |
Plastic Box |
2°C to 8°C | |
United States | |
Growth Promotion Testing, Pharmaceutical Microbial Enumeration Testing, Sterility Testing | |
5 Pellets |
Safety and Handling
ShelfLife : 8 Months (Minimum)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title