Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056358 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056358 0.1 mg
NB10056358T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056358 Supplier Novus Biologicals Supplier No. NB10056358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

EZH1 Polyclonal specifically detects EZH1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen EZH1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 4 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q92800
Gene Alias EC 2.1.1.43, Enhancer of zeste homolog 1, enhancer of zeste homolog 1 (Drosophila), ENX-2, histone-lysine N-methyltransferase EZH1, KIAA0388enhancer of zeste (Drosophila) homolog 1, KMT6B
Gene Symbols EZH1
Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human EZH1 was used as the immunogen.
Molecular Weight of Antigen 85.3 kDa
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 2145
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 93% homologous in mouse and 86% homologous in rat and dog.
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.