Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

$206.50 - $499.50

Specifications

Antigen EZH1
Concentration 1 mg/mL
Dilution Western Blot 2 - 4 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056358T
SDP
View Documents
Novus Biologicals
NB10056358SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056358
SDP
View Documents
Novus Biologicals
NB10056358
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

EZH1 Polyclonal specifically detects EZH1 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

EZH1
Western Blot 2 - 4 ug/ml
Polyclonal
Purified
RUO
Human, Mouse
Q92800
2145
A portion of amino acids 350-400 of human EZH1 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
85.3 kDa
1 mg/mL
Western Blot
Unconjugated
Rabbit
Chromatin Research
PBS with 0.05% Sodium Azide
EC 2.1.1.43, Enhancer of zeste homolog 1, enhancer of zeste homolog 1 (Drosophila), ENX-2, histone-lysine N-methyltransferase EZH1, KIAA0388enhancer of zeste (Drosophila) homolog 1, KMT6B
EZH1
IgG
Protein G purified
The amino acid sequence used as immunogen is 100% homologous in human, 93% homologous in mouse and 86% homologous in rat and dog.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.