Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH2/KMT6 Antibody (2C7), Novus Biologicals™
SDP

Catalog No. NB297781 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297781 100 μL
NB297780 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297781 Supplier Novus Biologicals Supplier No. NBP326598100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EZH2/KMT6 Monoclonal antibody specifically detects EZH2/KMT6 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen EZH2/KMT6
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 2C7
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169
Host Species Rabbit
Immunogen A synthesized peptide derived from Human EZH2/KMT6 [UniProt Q15910]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Phospho Specific, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2146
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.