Learn More
Description
Specifications
Specifications
| Antigen | EZH2/KMT6 |
| Applications | ChIP Assay |
| Classification | Monoclonal |
| Clone | C.15200180 |
| Conjugate | Unconjugated |
| Dilution | Chromatin Immunoprecipitation (ChIP) 2.5 ug |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169 |
| Gene Symbols | EZH2 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
