Learn More
Description
Specifications
Specifications
| Antigen | EZH2/KMT6 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, ChIP Assay, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP) 5 ug/IP, Chromatin Immunoprecipitation Sequencing, Knockdown Validated |
| Formulation | PBS with 0.05% Sodium Azide and 0.05% ProClin 300 |
| Gene Alias | EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169 |
| Gene Symbols | EZH2 |
| Host Species | Rabbit |
| Immunogen | The exact immunogen is proprietary information. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
