Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH2/KMT6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP217087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP217087 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP217087 Supplier Novus Biologicals Supplier No. NBP217087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EZH2/KMT6 Polyclonal specifically detects EZH2/KMT6 in Human, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen EZH2/KMT6
Applications Western Blot, Immunocytochemistry, Immunofluorescence, ChIP Assay
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Chromatin Immunoprecipitation (ChIP)
Formulation 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q15910
Gene Alias EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169
Gene Symbols EZH2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EZH2. The exact sequence is proprietary.
Molecular Weight of Antigen 85 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2146
Test Specificity Human. Expected to cross react based on sequence identity: Bovine (100%), Rhesus Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.