Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ezrin Antibody (2086A), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB374919 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374919 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374919 Supplier Novus Biologicals Supplier No. FAB7239I
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ezrin Monoclonal antibody specifically detects Ezrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry

Specifications

Antigen Ezrin
Applications Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Classification Monoclonal
Clone 2086A
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2
Host Species Rabbit
Immunogen E. coli-derived recombinant human Ezrin, Lys438-Arg562, Accession # P15311
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7430
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.