Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ezrin Antibody (6F1A9) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252465 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252465 Supplier Novus Biologicals Supplier No. NBP252465
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ezrin Monoclonal specifically detects Ezrin in Human, Mouse, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.

Specifications

Antigen Ezrin
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone 6F1A9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:400, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2
Gene Symbols EZR
Host Species Mouse
Immunogen Purified recombinant fragment of human EZR (AA: 292-464) expressed in E. Coli.
Molecular Weight of Antigen 69.4 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7430
Target Species Human, Mouse, Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.