Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ezrin Antibody (CL2384), Novus Biologicals™
SDP

Catalog No. NBP236760 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236760 0.1 mL
NB397080 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP236760 Supplier Novus Biologicals Supplier No. NBP236760
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ezrin Monoclonal specifically detects Ezrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Ezrin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL2384
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P15311
Gene Alias CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2
Gene Symbols EZR
Host Species Mouse
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:REEKHQKQLERQQLETEKKRRETVEREKEQMMREKEELMLRLQDYEEKTKKAERELSEQIQRALQLEEERKRAQEEAERLEADRMAALRAKEELERQAVDQIKSQEQLAAELAEYTAKIALLE
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7430
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Reconstitution Protein A purified
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.