Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ezrin/Radixin/Moesin, p Thr567, p Thr564, p Thr558 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB168995 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168995 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB168995 Supplier Novus Biologicals Supplier No. NBP316959
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ezrin/Radixin/Moesin Polyclonal antibody specifically detects Ezrin/Radixin/Moesin in Human samples. It is validated for Western Blot

Specifications

Antigen Ezrin/Radixin/Moesin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias CVIL, CVL, Cytovillin, DFNB24, EZR, HEL70, HEL-S-105, IMD50, Membrane-organizing extension spike protein, MSN, p81, RDX, villin-2
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around T567 of human Ezrin/Radixin/Moesin (NP_003370.2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7430
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.