Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-box protein 15/FBXO15 Antibody (612834), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB295960 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295960 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295960 Supplier Novus Biologicals Supplier No. FAB6035S

Mouse Monoclonal Antibody

F-box protein 15/FBXO15 Monoclonal antibody specifically detects F-box protein 15/FBXO15 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen F-box protein 15/FBXO15
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 612834
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias F-box only protein 15, F-box protein 15, FBX15MGC39671
Host Species Mouse
Immunogen E. coli-derived recombinant human F-box protein 15/FBXO15 isoform 2,, Asn298-Tyr434, Accession # Q8NCQ5-2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201456
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.