Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F4/80 Antibody (521204), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB311988 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311988 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311988 Supplier Novus Biologicals Supplier No. FAB5580APCCY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen F4/80
Applications Flow Cytometry
Classification Monoclonal
Clone 521204
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias ADGRE1, cell surface glycoprotein F4/80, DD7A5-, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, Emr, Emr1, EMR1 hormone receptor, F4/80, Gpf480, Ly7, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7L, TM7LN3
Host Species Rat
Immunogen NS0 mouse myeloma cell line transfected with mouse F4/80/EMR1, Gln28-Gly931, Accession # Q61549
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cellular Markers, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 13733
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.