Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F4/80 Antibody (CI-A3-1), Biotin, Novus Biologicals™
SDP

Catalog No. NB211346 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211346 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211346 Supplier Novus Biologicals Supplier No. NB600404B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

F4/80 Monoclonal antibody specifically detects F4/80 in Human, Mouse, C. elegans, E. coli samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunomicroscopy

Specifications

Antigen F4/80
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunomicroscopy
Classification Monoclonal
Clone CI-A3-1
Conjugate Biotin
Formulation PBS
Gene Alias ADGRE1, cell surface glycoprotein F4/80, DD7A5-, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, Emr, Emr1, EMR1 hormone receptor, F4/80, Gpf480, Ly7, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7L, TM7LN3
Host Species Rat
Immunogen This F4/80 Antibody (CI-A3-1) was developed against Thioglycollate stimulated peritoneal macrophages from C57BL/6 mice.
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cellular Markers, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 13733
Target Species Human, Mouse, C. elegans, E. coli
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.