Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Epitope Tag Antibody (L5), Novus Biologicals™
SDP

Catalog No. NBP106712 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106712 0.5 mL
NBP106712SS 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP106712 Supplier Novus Biologicals Supplier No. NBP106712
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 75 publications

DYKDDDDK Epitope Tag Monoclonal specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen).

Specifications

Antigen DYKDDDDK Epitope Tag
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone L5
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000. Use reported in multiple pieces of scientific literature, Immunohistochemistry 1:50-1:100. Use reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:100. Use reported in multiple pieces of scientific literature, Immunoprecipitation reported in multiple pieces of scientific literature, Immunohistochemistry-Frozen reported in scientific literature (PMID 24454782)
Formulation Tissue culture supernatant with 0.1% Sodium Azide
Gene Alias anti-flag L5, DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
Host Species Rat
Immunogen DYKDDDDK Epitope Tag Antibody (L5) was made to N-terminal DYKDDDDK-tagged extracellular domain of mouse Langerin. Binds to same epitope as SigmA′s Anti-FLAG® M2 Antibody.
Molecular Weight of Antigen 1.01 kDa
Purification Method Tissue culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline ECS Epitope Tags (DDDDK), Epitope Tags
Primary or Secondary Primary
Test Specificity Binds to same epitope as Sigma's Anti-FLAGM2 Antibody. FLAG is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
Target Species Guinea Pig
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgG2a
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.