Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYKDDDDK Epitope Tag Antibody (L5), Novus Biologicals™

Rat Monoclonal Antibody has been used in 75 publications
$254.00 - $606.50
Specifications
Antigen | DYKDDDDK Epitope Tag |
---|---|
Clone | L5 |
Dilution | Western Blot 1:500-1:1000. Use reported in multiple pieces of scientific literature, Immunohistochemistry 1:50-1:100. Use reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:100. Use reported in multiple pieces of scientific literature, Immunoprecipitation reported in multiple pieces of scientific literature, Immunohistochemistry-Frozen reported in scientific literature (PMID 24454782) |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP106712SS
![]() |
Novus Biologicals
NBP106712SS |
0.125 mL |
Each for $254.00
|
|
|||||
NBP106712
![]() |
Novus Biologicals
NBP106712 |
0.5 mL |
Each for $606.50
|
|
|||||
Description
DYKDDDDK Epitope Tag Monoclonal specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen).Specifications
DYKDDDDK Epitope Tag | |
Western Blot 1:500-1:1000. Use reported in multiple pieces of scientific literature, Immunohistochemistry 1:50-1:100. Use reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:100. Use reported in multiple pieces of scientific literature, Immunoprecipitation reported in multiple pieces of scientific literature, Immunohistochemistry-Frozen reported in scientific literature (PMID 24454782) | |
Monoclonal | |
Supernatant | |
RUO | |
Guinea Pig | |
anti-flag L5, DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag | |
IgG2a | |
Tissue culture supernatant | |
Binds to same epitope as Sigma's Anti-FLAGM2 Antibody. FLAG is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. |
L5 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
Unconjugated | |
Rat | |
ECS Epitope Tags (DDDDK), Epitope Tags | |
Tissue culture supernatant with 0.1% Sodium Azide | |
DYKDDDDK Epitope Tag Antibody (L5) was made to N-terminal DYKDDDDK-tagged extracellular domain of mouse Langerin. Binds to same epitope as SigmA′s Anti-FLAG® M2 Antibody. | |
Primary | |
Store at 4C. Do not freeze. | |
1.01 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title