Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F4/80 Antibody (CI-A3-1), PerCP, Novus Biologicals™
SDP

Catalog No. NB600404PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600404PCP 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600404PCP Supplier Novus Biologicals Supplier No. NB600404PCP
Add to Cart
Add to Cart

Rat Monoclonal Antibody

F4/80 Monoclonal specifically detects F4/80 in Human, Mouse, C. elegans, E. coli samples. It is validated for Flow Cytometry.

Specifications

Antigen F4/80
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone CI-A3-1
Conjugate PerCP
Dilution Flow Cytometry
Gene Alias ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3
Gene Symbols ADGRE1
Host Species Rat
Immunogen Thioglycollate stimulated peritoneal macrophages from C57BL/6 mice.
Molecular Weight of Antigen 160 kDa
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 2015
Test Specificity This recognizes mouse F4/80, a 160kD glycoprotein expressed by murine macrophages.
Target Species Human, Mouse, C. elegans, E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.