Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F4/80 Antibody (Cl:A3-1 (recombinant version)), PE, Novus Biologicals™
SDP

Catalog No. NB014374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014374 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014374 Supplier Novus Biologicals Supplier No. NBP281029PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

F4/80 Monoclonal specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen F4/80
Applications Flow Cytometry
Classification Monoclonal
Clone Cl:A3-1 (recombinant version)
Conjugate PE
Dilution Flow Cytometry
Formulation PBS
Gene Alias ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3
Gene Symbols Adgre1
Host Species Rabbit
Immunogen Thioglycollate stimulated peritoneal macrophages of mouse origin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, GPCR, Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 13733
Test Specificity This antibody recognises the mouse F4/80 antigen, a 160kD glycoprotein, expressed by murine macrophages.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.