Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FA2H Antibody (OTI2E3), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP272192UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272192UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272192UV Supplier Novus Biologicals Supplier No. NBP272192UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FA2H Monoclonal specifically detects FA2H in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FA2H
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2E3
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FAAHFAXDC1, FAH1, fatty acid 2-hydroxylase, Fatty acid alpha-hydroxylase, fatty acid hydroxylase domain containing 1, FLJ25287, SCS7, spastic paraplegia 35 (autosomal recessive), SPG35
Gene Symbols FA2H
Host Species Mouse
Immunogen Full length recombinant protein of human FA2H (NP_077282) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 79152.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.