Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FA2H Antibody (OTI2E3), PE, Novus Biologicals™
SDP

Catalog No. NB288329 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288329 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288329 Supplier Novus Biologicals Supplier No. NBP272192PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FA2H Monoclonal antibody specifically detects FA2H in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen FA2H
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2E3
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias FAAHFAXDC1, FAH1, fatty acid 2-hydroxylase, Fatty acid alpha-hydroxylase, fatty acid hydroxylase domain containing 1, FLJ25287, SCS7, spastic paraplegia 35 (autosomal recessive), SPG35
Host Species Mouse
Immunogen Full length human recombinant protein of human FA2H(NP_077282) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 79152
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.