Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FA2H Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093860 20 μL
NB093859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093860 Supplier Novus Biologicals Supplier No. NBP30518620UL

Rabbit Polyclonal Antibody

FA2H Polyclonal specifically detects FA2H in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FA2H
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias FAAHFAXDC1, FAH1, fatty acid 2-hydroxylase, Fatty acid alpha-hydroxylase, fatty acid hydroxylase domain containing 1, FLJ25287, SCS7, spastic paraplegia 35 (autosomal recessive), SPG35
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 95-170 of human FA2H (NP_077282.3).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 79152
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.