Learn More
Description
Specifications
Specifications
| Antigen | FAAP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | ankyrin repeat domain 54, C22orf28, chromosome 22 open reading frame 28, DJ149A16.6, EC 6.5.1.3, HSPC117, hypothetical protein LOC51493, novel protein HSPC117, RP1-149A16.6 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 236-505 of human FAAP (NP_055121.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
