Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FAAP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FAAP Polyclonal specifically detects FAAP in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
FAAP | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
51493 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
ankyrin repeat domain 54, C22orf28, chromosome 22 open reading frame 28, DJ149A16.6, EC 6.5.1.3, HSPC117, hypothetical protein LOC51493, novel protein HSPC117, RP1-149A16.6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 236-505 of human FAAP (NP_055121.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title